The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293108.1 | NP_001280037.1 | - | 994 | 80 |
NM_001293109.1 | NP_001280038.1 | - | 869 | 80 | |
XM_015298897.2 | XP_015154383.1 | - | 1074 | 80 | |
XM_025143679.1 | XP_024999447.1 | - | 948 | 80 | |
GenBank mRNA | BU313448.1 | - | - | 446 | 80 |
BU337330.1 | - | - | 482 | 80 | |
M26810.1 | - | - | 252 | 80 |