Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARL10
Entrez Gene ID
416229
Gene Name
ADP ribosylation factor like GTPase 10
Gene Aliases
ARL10A
Chromosome Location
Chr.13: 11124638 - 11127201 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277714.1 NP_001264643.1 2 - 3 378 88
GenBank mRNA BX929782.1 - 2 - 3 378 88
BX933616.1 - 2 - 3 396 88

View Full Product Details