Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPDL1
Entrez Gene ID
416172
Gene Name
spindle apparatus coiled-coil protein 1
Gene Aliases
CCDC99
Chromosome Location
Chr.13: 4874941 - 4897864 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004944787.3 XP_004944844.2 1 - 2 48 147
XM_025154966.1 XP_025010734.1 1 - 2 48 157
XM_025154967.1 XP_025010735.1 1 - 2 47 147
XM_414505.6 XP_414505.3 1 - 2 48 157
GenBank mRNA BX930082.1 - 1 - 2 37 157
BX936052.1 - 1 - 2 61 145
BX936106.1 - 1 - 2 61 145

View Full Product Details