Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NINJ1
Entrez Gene ID
415986
Gene Name
ninjurin 1
Gene Aliases
-
Chromosome Location
Chr.12: 7012303 - 7029920 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BX930736.1 - 3 - 4 594 67

View Full Product Details