Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSBP1L1
Entrez Gene ID
420812
Gene Name
heat shock factor binding protein 1 like 1
Gene Aliases
-
Chromosome Location
Chr.2: 56182363 - 56185362 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136182.2 NP_001129654.1 3 - 4 407 83
GenBank mRNA BX932365.1 - 3 - 4 393 83
BX934295.2 - 3 - 4 288 83
DN853252.1 - 3 - 4 407 83

View Full Product Details