Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALKBH2
Entrez Gene ID
416897
Gene Name
alkB homolog 2, alpha-ketoglutarate dependent dioxygenase
Gene Aliases
-
Chromosome Location
Chr.15: 6944042 - 6946525 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277499.1 NP_001264428.1 1 - 2 332 83
GenBank mRNA BX932990.1 - 1 - 2 339 83

View Full Product Details