Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAUS2
Entrez Gene ID
423237
Gene Name
HAUS augmin like complex subunit 2
Gene Aliases
CEP27
Chromosome Location
Chr.5: 25961526 - 25966227 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199667.1 NP_001186596.1 4 - 5 525 69
XM_015287080.2 XP_015142566.1 5 - 6 563 69
GenBank mRNA BX933538.1 - 4 - 5 525 69

View Full Product Details