Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDHAF3
Entrez Gene ID
420572
Gene Name
succinate dehydrogenase complex assembly factor 3
Gene Aliases
ACN9
Chromosome Location
Chr.2: 24478719 - 24505947 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_418673.5 XP_418673.1 1 - 2 215 125
GenBank mRNA BX934538.1 - 1 - 2 193 125

View Full Product Details