Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TCTEX1D2
- Entrez Gene ID
-
424914
- Gene Name
- Tctex1 domain containing 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 15052970 - 15057284 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_422724.5 |
XP_422724.1 |
4 - 5 |
387 |
77 |
GenBank mRNA |
BX932411.2 |
- |
4 - 5 |
406 |
77 |
- Gene Symbol
- PCYT1A
- Entrez Gene ID
-
424915
- Gene Name
- phosphate cytidylyltransferase 1, choline, alpha
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 15053043 - 15076424 on Build GRCg6a
View Full Product Details