Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAF7
Entrez Gene ID
422192
Gene Name
TATA-box binding protein associated factor 7
Gene Aliases
TAF7L
Chromosome Location
Chr.4: 2044739 - 2051320 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277886.1 NP_001264815.1 10 - 11 1049 77
XM_015278223.2 XP_015133709.1 10 - 11 1064 77
XM_015278224.2 XP_015133710.1 9 - 10 1149 77
XM_025149547.1 XP_025005315.1 9 - 10 1149 77
GenBank mRNA AJ444344.1 - 2 - 3 378 77
BX930950.1 - 10 - 11 1035 77
BX935900.2 - 9 - 10 1135 77

View Full Product Details