Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHOX2B
Entrez Gene ID
395548
Gene Name
paired like homeobox 2b
Gene Aliases
-
Chromosome Location
Chr.4: 68436612 - 68440559 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015285470.2 XP_015140956.1 1 - 2 2269 97
GenBank mRNA AJ251458.1 - 1 - 2 179 97

View Full Product Details