Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHST15
Entrez Gene ID
423952
Gene Name
carbohydrate sulfotransferase 15
Gene Aliases
GALNAC4S-6ST
Chromosome Location
Chr.6: 32816682 - 32859776 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_421811.6 XP_421811.2 1 - 2 86 112
GenBank mRNA BX929349.1 - 1 - 2 82 112

View Full Product Details