The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_004947398.3 | XP_004947455.1 | 6 - 7 | 2802 | 60 |
XM_025142634.1 | XP_024998402.1 | 6 - 7 | 820 | 60 | |
XM_417569.4 | XP_417569.1 | 6 - 7 | 1303 | 60 | |
GenBank mRNA | BX929570.1 | - | 6 - 7 | 747 | 60 |
BX934969.1 | - | 6 - 7 | 892 | 60 |