Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERICH1
Entrez Gene ID
421904
Gene Name
glutamate rich 1
Gene Aliases
-
Chromosome Location
Chr.3: 91475835 - 91569878 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278029.1 NP_001264958.1 5 - 6 995 133
GenBank mRNA BX930034.1 - 5 - 6 958 133

View Full Product Details