Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RTKN2
Entrez Gene ID
423660
Gene Name
rhotekin 2
Gene Aliases
PLEKHK1, rhotekin-2
Chromosome Location
Chr.6: 8782767 - 8954175 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199597.1 NP_001186526.1 11 - 12 1191 90
XM_015288109.2 XP_015143595.1 10 - 11 1210 90
XM_025151516.1 XP_025007284.1 11 - 12 1291 90
XM_025151517.1 XP_025007285.1 11 - 12 1291 90
XM_025151518.1 XP_025007286.1 11 - 12 1291 90
XM_025151519.1 XP_025007287.1 11 - 12 1291 90
XM_025151520.1 XP_025007288.1 11 - 12 1402 90
XM_025151521.1 XP_025007289.1 23 - 24 4509 90
XM_025151522.1 XP_025007290.1 23 - 24 12607 90
XM_025151523.1 XP_025007291.1 23 - 24 4395 90
XM_025151524.1 XP_025007292.1 24 - 25 11644 90
XM_025151525.1 XP_025007293.1 24 - 25 12741 90
XM_025151526.1 XP_025007294.1 23 - 24 12501 90
XM_025151527.1 XP_025007295.1 24 - 25 11597 90
XM_025151528.1 XP_025007296.1 22 - 23 12419 90
XM_025151529.1 XP_025007297.1 23 - 24 11537 90
XM_025151530.1 XP_025007298.1 23 - 24 3285 90
XM_025151531.1 XP_025007299.1 22 - 23 12181 90
XM_025151532.1 XP_025007300.1 24 - 25 12698 90
XM_025151533.1 XP_025007301.1 22 - 23 12548 90
XM_025151534.1 XP_025007302.1 21 - 22 3050 90
XM_025151535.1 XP_025007303.1 16 - 17 11252 90
XM_025151536.1 XP_025007304.1 13 - 14 2911 90
XM_025151537.1 XP_025007305.1 13 - 14 2048 90
XM_025151538.1 XP_025007306.1 14 - 15 5207 90
XM_025151539.1 XP_025007307.1 12 - 13 1627 90
XM_025151540.1 XP_025007308.1 13 - 14 3633 90
XM_025151541.1 XP_025007309.1 13 - 14 3614 90
XM_025151543.1 XP_025007311.1 10 - 11 1210 90
XM_025151544.1 XP_025007312.1 22 - 23 12566 90
XM_025151545.1 XP_025007313.1 10 - 11 1090 90
GenBank mRNA CR391233.1 - 4 - 5 504 90

View Full Product Details