Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFB6
Entrez Gene ID
416391
Gene Name
NADH:ubiquinone oxidoreductase subunit B6
Gene Aliases
-
Chromosome Location
Chr.14: 13797513 - 13799635 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277525.1 NP_001264454.1 1 - 2 263 102
GenBank mRNA BX930426.1 - 1 - 2 263 102

View Full Product Details