Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GSTK1
Entrez Gene ID
418302
Gene Name
glutathione S-transferase kappa 1
Gene Aliases
-
Chromosome Location
Chr.1: 78425551 - 78436579 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198649.1 NP_001185578.1 5 - 6 467 72
GenBank mRNA BX930797.1 - 5 - 6 431 72
BX931011.2 - 5 - 6 467 72

View Full Product Details