Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PBLD
Entrez Gene ID
423685
Gene Name
phenazine biosynthesis like protein domain containing
Gene Aliases
-
Chromosome Location
Chr.6: 11263100 - 11281943 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277337.1 NP_001264266.1 7 - 8 883 69
XM_015278680.2 XP_015134166.1 7 - 8 742 69
GenBank mRNA BU307502.1 - 1 - 2 194 69
BX934609.1 - 7 - 8 761 69

View Full Product Details