Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCLAF1
Entrez Gene ID
100858086
Gene Name
BCL2-associated transcription factor 1
Gene Aliases
-
Chromosome Location
Chr.3: 55290217 - 55317813 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004935711.3 XP_004935768.2 6 - 7 2080 66
XM_004935712.2 XP_004935769.2 5 - 6 1947 66
XM_004935713.3 XP_004935770.2 5 - 6 1940 66
XM_015284178.2 XP_015139664.1 6 - 7 2079 66
XM_015284179.2 XP_015139665.1 5 - 6 1911 66
XM_015284180.2 XP_015139666.1 6 - 7 2081 66
XM_025149344.1 XP_025005112.1 5 - 6 1325 66
GenBank mRNA CR352654.1 - 1 - 2 435 66

View Full Product Details