Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GDPD4
Entrez Gene ID
419084
Gene Name
glycerophosphodiester phosphodiesterase domain containing 4
Gene Aliases
-
Chromosome Location
Chr.1: 194560292 - 194594332 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015280883.2 XP_015136369.1 2 - 3 369 73
XM_417276.6 XP_417276.2 2 - 3 300 73
GenBank mRNA BX935679.2 - 2 - 3 286 73

View Full Product Details