Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEX30
Entrez Gene ID
418767
Gene Name
testis expressed 30
Gene Aliases
C13orf27, C1H13ORF27
Chromosome Location
Chr.1: 144242942 - 144249023 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277762.1 NP_001264691.1 4 - 5 593 67
GenBank mRNA BU297448.1 - 4 - 5 593 67
BX950616.1 - 4 - 5 593 67

View Full Product Details