Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSBP1
Entrez Gene ID
418122
Gene Name
single stranded DNA binding protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 57338774 - 57344343 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278007.1 NP_001264936.1 4 - 5 417 105
GenBank mRNA BU286106.1 - 4 - 5 368 105
BX932437.1 - 4 - 5 414 105
CR352496.1 - 4 - 5 388 105

View Full Product Details