Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EAPP
Entrez Gene ID
423319
Gene Name
E2F associated phosphoprotein
Gene Aliases
-
Chromosome Location
Chr.5: 36120589 - 36125843 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197245.1 NP_001184174.1 5 - 6 629 75
GenBank mRNA BX935038.2 - 5 - 6 629 75

View Full Product Details