Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PACRGL
Entrez Gene ID
422818
Gene Name
parkin coregulated like
Gene Aliases
C4orf28
Chromosome Location
Chr.4: 74923536 - 74962589 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025149901.1 XP_025005669.1 2 - 3 409 131
XM_025149902.1 XP_025005670.1 2 - 3 409 131
XM_025149903.1 XP_025005671.1 2 - 3 409 131
XM_025149904.1 XP_025005672.1 2 - 3 409 131
XM_025149906.1 XP_025005674.1 2 - 3 409 131
XM_025149907.1 XP_025005675.1 2 - 3 409 131
XM_025149908.1 XP_025005676.1 2 - 3 409 131
XM_025149909.1 XP_025005677.1 2 - 3 409 131
XM_420764.6 XP_420764.2 2 - 3 409 131
GenBank mRNA BX930771.1 - 2 - 3 207 131

View Full Product Details