Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHDC8A
Entrez Gene ID
419829
Gene Name
kelch domain containing 8A
Gene Aliases
-
Chromosome Location
Chr.26: 2087356 - 2099662 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015299083.2 XP_015154569.1 - 1444 75
XM_015299084.2 XP_015154570.1 - 1451 75
XM_015299085.2 XP_015154571.1 - 1546 75
XM_015299086.2 XP_015154572.1 - 1201 75
XM_417963.6 XP_417963.3 - 1421 75
GenBank mRNA BX935746.1 - - 852 75

View Full Product Details