Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINB6
Entrez Gene ID
420895
Gene Name
serpin peptidase inhibitor, clade B (ovalbumin), member 6
Gene Aliases
SERPINB6L
Chromosome Location
Chr.2: 67630807 - 67640267 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006377.1 NP_001006377.1 3 - 4 312 146
XM_015282170.2 XP_015137656.1 3 - 4 398 146
XM_025147268.1 XP_025003036.1 3 - 4 359 146
GenBank mRNA AJ719818.1 - 3 - 4 312 146

View Full Product Details