Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO8
Entrez Gene ID
422565
Gene Name
F-box protein 8
Gene Aliases
-
Chromosome Location
Chr.4: 43677976 - 43697009 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006446.1 NP_001006446.1 3 - 4 662 77
XM_015276238.2 XP_015131724.1 3 - 4 665 77
GenBank mRNA AJ720952.1 - 3 - 4 662 77

View Full Product Details