Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NR2E3
Entrez Gene ID
395289
Gene Name
nuclear receptor subfamily 2 group E member 3
Gene Aliases
PNR
Chromosome Location
Chr.10: 1852294 - 1857167 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204594.1 NP_989925.1 5 - 6 764 88
XM_025153796.1 XP_025009564.1 4 - 5 794 88
GenBank mRNA AY053465.1 - 5 - 6 764 88

View Full Product Details