Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HABP4
Entrez Gene ID
395444
Gene Name
hyaluronan binding protein 4
Gene Aliases
-
Chromosome Location
Chr.Z: 42191322 - 42213072 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204705.1 NP_990036.1 6 - 7 1180 74
XM_015280241.2 XP_015135727.1 6 - 7 1217 74
GenBank mRNA AF227683.1 - 6 - 7 1180 74

View Full Product Details