Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NTM
Entrez Gene ID
395450
Gene Name
neurotrimin
Gene Aliases
CEPU, CEPU-SE, HNT
Chromosome Location
Chr.24: 1826279 - 2138596 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204711.1 NP_990042.1 7 - 8 1022 60
XM_015297952.2 XP_015153438.1 7 - 8 1297 60
XM_015297955.2 XP_015153441.1 8 - 9 1169 60
XM_015297956.2 XP_015153442.1 7 - 8 891 60
XM_025143176.1 XP_024998944.1 6 - 7 821 60
GenBank mRNA Z72497.1 - 7 - 8 1022 60

View Full Product Details