Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
VAX1
Entrez Gene ID
395582
Gene Name
ventral anterior homeobox 1
Gene Aliases
cVAX
Chromosome Location
Chr.6: 30032849 - 30038215 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204799.2 NP_990130.2 2 - 3 425 60
XM_025151484.1 XP_025007252.1 3 - 4 442 60
GenBank mRNA AB032248.1 - 2 - 3 425 60
AF117299.1 - 2 - 3 461 60

View Full Product Details