Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEF
Entrez Gene ID
396342
Gene Name
TEF, PAR bZIP transcription factor
Gene Aliases
VBP
Chromosome Location
Chr.1: 49664271 - 49678819 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205382.1 NP_990713.1 4 - 5 985 96
GenBank mRNA U09221.1 - 4 - 5 985 96

View Full Product Details