Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YBX3
Entrez Gene ID
396371
Gene Name
Y-box binding protein 3
Gene Aliases
CSDA
Chromosome Location
Chr.1: 79075430 - 79099953 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205406.2 NP_990737.2 5 - 6 603 95
GenBank mRNA CD218569.1 - 5 - 6 603 95
U23162.1 - 5 - 6 592 95

View Full Product Details