Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANOS1
Entrez Gene ID
396395
Gene Name
anosmin 1
Gene Aliases
KAL, KAL1, anosmin-1
Chromosome Location
Chr.1: 127043959 - 127168215 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205424.1 NP_990755.1 11 - 12 1604 96
GenBank mRNA CR386644.1 - 8 - 9 1127 96
L12144.1 - 11 - 12 1604 96

View Full Product Details