Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPS15
Entrez Gene ID
396448
Gene Name
ribosomal protein S15
Gene Aliases
-
Chromosome Location
Chr.28: 3300740 - 3302010 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205462.1 NP_990793.1 1 - 2 91 83
GenBank mRNA M33331.1 - 1 - 2 91 83

View Full Product Details