Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLP1
Entrez Gene ID
423131
Gene Name
cleavage and polyadenylation factor I subunit 1
Gene Aliases
-
Chromosome Location
Chr.5: 17249282 - 17251526 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012292.1 NP_001012292.1 1 - 2 736 73
GenBank mRNA AJ720420.1 - 1 - 2 736 73

View Full Product Details