Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYLIP
Entrez Gene ID
420841
Gene Name
myosin regulatory light chain interacting protein
Gene Aliases
-
Chromosome Location
Chr.2: 61479711 - 61493936 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012561.1 NP_001012579.1 5 - 6 963 119
GenBank mRNA AJ851471.1 - 5 - 6 963 119
CR390454.1 - 4 - 5 563 119

View Full Product Details