Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC59
Entrez Gene ID
422103
Gene Name
leucine rich repeat containing 59
Gene Aliases
-
Chromosome Location
Chr.18: 10377548 - 10381847 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012570.1 NP_001012588.1 2 - 3 197 68
XM_015295391.2 XP_015150877.1 2 - 3 201 68
GenBank mRNA AJ851816.1 - 2 - 3 197 68
BX929298.1 - 2 - 3 180 68
BX929381.1 - 2 - 3 193 68
BX933636.2 - 2 - 3 187 68

View Full Product Details