Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTMR8
Entrez Gene ID
422312
Gene Name
myotubularin related protein 8
Gene Aliases
-
Chromosome Location
Chr.4: 11625916 - 11649930 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012699.1 NP_001012717.1 11 - 12 1445 65
GenBank mRNA AJ851448.1 - 11 - 12 1445 65

View Full Product Details