Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBFA
Entrez Gene ID
420806
Gene Name
ribosome binding factor A (putative)
Gene Aliases
C18orf22
Chromosome Location
Chr.2: 56089456 - 56105012 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012861.1 NP_001012879.1 1 - 2 174 113
GenBank mRNA AJ719502.1 - 1 - 2 174 113

View Full Product Details