Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WIPF1
Entrez Gene ID
424143
Gene Name
WAS/WASL interacting protein family member 1
Gene Aliases
WASPIP
Chromosome Location
Chr.7: 16811407 - 16865355 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012917.1 NP_001012935.1 4 - 5 366 155
XM_015289441.2 XP_015144927.2 4 - 5 547 155
XM_015289442.2 XP_015144928.2 3 - 4 397 155
XM_015289443.2 XP_015144929.2 3 - 4 283 155
XM_015289444.2 XP_015144930.2 3 - 4 411 155
XM_015289445.2 XP_015144931.2 3 - 4 334 155
XM_025152191.1 XP_025007959.1 4 - 5 826 155
GenBank mRNA AJ851593.1 - 4 - 5 366 155

View Full Product Details