Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMAD5
Entrez Gene ID
395679
Gene Name
SMAD family member 5
Gene Aliases
-
Chromosome Location
Chr.13: 15806654 - 15833246 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014968.1 NP_001014968.1 1 - 2 419 100
XM_015293967.2 XP_015149453.1 2 - 3 729 100
XM_015293968.2 XP_015149454.1 2 - 3 651 100
XM_025154805.1 XP_025010573.1 3 - 4 3664 100
GenBank mRNA AF143240.1 - 1 - 2 306 100
AY953144.1 - 1 - 2 419 100
BX932577.2 - 2 - 3 677 100

View Full Product Details