Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL21
Entrez Gene ID
554218
Gene Name
interleukin 21
Gene Aliases
interleukin-21
Chromosome Location
Chr.4: 53679705 - 53684460 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024835.1 NP_001020006.1 4 - 5 382 68
GenBank mRNA AM773757.1 - 4 - 5 382 68
AY943833.1 - 4 - 5 382 68
FJ788638.1 - 4 - 5 382 68

View Full Product Details