Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFDP1
Entrez Gene ID
374073
Gene Name
craniofacial development protein 1
Gene Aliases
bcnt
Chromosome Location
Chr.11: 1980932 - 2031882 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001189.1 NP_001001189.1 5 - 6 667 74
GenBank mRNA AB080800.1 - 3 - 4 420 74
AB162146.1 - 5 - 6 667 74

View Full Product Details