Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRT13
Entrez Gene ID
408040
Gene Name
keratin 13
Gene Aliases
KRT15
Chromosome Location
Chr.27: 7361940 - 7366365 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001312.2 NP_001001312.2 7 - 8 1363 138
GenBank mRNA AY574986.1 - 8 - 9 1265 138
BU389884.1 - 3 - 4 298 138
CN227144.1 - 5 - 6 625 138

View Full Product Details