Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST8SIA5
Entrez Gene ID
414797
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 5
Gene Aliases
SIAT8E
Chromosome Location
Chr.Z: 1821762 - 1881537 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001747.2 NP_001001747.2 6 - 7 1292 108
XM_004949029.3 XP_004949086.1 7 - 8 1330 108
XM_004949031.3 XP_004949088.1 5 - 6 4645 108
XM_004949032.3 XP_004949089.1 5 - 6 1128 108
XM_015277412.2 XP_015132898.1 6 - 7 4737 108
XM_015277413.2 XP_015132899.1 5 - 6 1082 108
XM_015277414.2 XP_015132900.1 5 - 6 1100 108
XM_015277415.2 XP_015132901.1 4 - 5 4537 108
XM_015277416.1 XP_015132902.1 4 - 5 318 108
GenBank mRNA AJ704564.1 - 2 - 3 318 108

View Full Product Details