Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RACK1
Entrez Gene ID
417044
Gene Name
receptor for activated C kinase 1
Gene Aliases
C12-3, GNB2L1, c12.3
Chromosome Location
Chr.16: 2526354 - 2529817 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004378.2 NP_001004378.1 4 - 5 624 78
GenBank mRNA AY393848.1 - 4 - 5 522 78
BU137661.1 - 3 - 4 367 78
CR386189.1 - 4 - 5 604 78
M24193.1 - 4 - 5 604 78

View Full Product Details