Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLBP
Entrez Gene ID
422902
Gene Name
stem-loop binding protein
Gene Aliases
-
Chromosome Location
Chr.4: 83930355 - 83937106 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004403.2 NP_001004403.1 2 - 3 264 83
GenBank mRNA AJ719644.1 - 2 - 3 264 83
AY675345.1 - 2 - 3 251 83

View Full Product Details