Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VTG1
Entrez Gene ID
424547
Gene Name
vitellogenin 1
Gene Aliases
LVII, TPRXL, VIT1, YGP42, vitellogenin-1
Chromosome Location
Chr.8: 18989399 - 19032035 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004408.2 NP_001004408.1 8 - 9 1107 71
GenBank mRNA D89547.1 - 8 - 9 1108 71

View Full Product Details