Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMC1
Entrez Gene ID
395804
Gene Name
proteasome 26S subunit, ATPase 1
Gene Aliases
-
Chromosome Location
Chr.5: 44001300 - 44009365 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204958.1 NP_990289.1 3 - 4 295 148
GenBank mRNA U60187.1 - 3 - 4 295 148

View Full Product Details